Product Details

SNP ID
rs116967618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62863884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGACTTGCATTGTGCTTTTGCC[A/G]TAGGAGCTTGCCATAAGTTCTACAC
Phenotype
MIM: 158070 MIM: 603222 MIM: 603224 MIM: 603225 MIM: 603226 MIM: 603227 MIM: 603228
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC3A2 PubMed Links

Gene Details

Gene
SLC3A2
Gene Name
solute carrier family 3 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012662.2 Intron NP_001012680.1
NM_001012664.2 Intron NP_001012682.1
NM_001013251.2 Intron NP_001013269.1
NM_002394.5 Intron NP_002385.3
Gene
SNHG1
Gene Name
small nucleolar RNA host gene 1
There are no transcripts associated with this gene.

Gene
SNORD25
Gene Name
small nucleolar RNA, C/D box 25
There are no transcripts associated with this gene.

Gene
SNORD26
Gene Name
small nucleolar RNA, C/D box 26
There are no transcripts associated with this gene.

Gene
SNORD27
Gene Name
small nucleolar RNA, C/D box 27
There are no transcripts associated with this gene.

Gene
SNORD28
Gene Name
small nucleolar RNA, C/D box 28
There are no transcripts associated with this gene.

Gene
SNORD29
Gene Name
small nucleolar RNA, C/D box 29
There are no transcripts associated with this gene.

View Full Product Details