Product Details

SNP ID
rs117284601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57657756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCACAGCGCAAGTGCGGAACT[A/G]CGTTTGTTTCCGGCGTGGGTCCGGG
Phenotype
MIM: 608757 MIM: 609725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLP1 PubMed Links

Gene Details

Gene
CLP1
Gene Name
cleavage and polyadenylation factor I subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142597.1 13 UTR 5 NP_001136069.1
NM_006831.2 13 UTR 5 NP_006822.1
Gene
YPEL4
Gene Name
yippee like 4
There are no transcripts associated with this gene.

View Full Product Details