Product Details
- SNP ID
-
rs117353413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:63554843 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAAAAGAAAGGGGGAAAAAAAA[C/T]CATTTTATTCAAATACACTCAGAAT
- Phenotype
-
MIM: 613866
MIM: 605092
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HRASLS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200768428] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HRASLS2
- Gene Name
- HRAS like suppressor 2
There are no transcripts associated with this gene.
- Gene
- RARRES3
- Gene Name
- retinoic acid receptor responder 3
There are no transcripts associated with this gene.
View Full Product Details