Product Details

SNP ID
rs118086344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57340604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCAGGCACTGTGGATTGAGGAC[A/G]GAATGGTGGGCAAAAGCAGACAGGG
Phenotype
MIM: 600843 MIM: 604328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
P2RX3 PubMed Links

Gene Details

Gene
P2RX3
Gene Name
purinergic receptor P2X 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002559.3 Intron NP_002550.2
XM_011545069.2 Intron XP_011543371.1
XM_011545070.2 Intron XP_011543372.1
XM_011545071.2 Intron XP_011543373.1
XM_011545072.2 Intron XP_011543374.1
XM_011545073.1 Intron XP_011543375.1
Gene
SSRP1
Gene Name
structure specific recognition protein 1
There are no transcripts associated with this gene.

View Full Product Details