Product Details

SNP ID
rs137996775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8696435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAACCAAGACTTACCCTTTGCC[C/T]GACACTTTCTTAGCTCCCTGTCTTG
Phenotype
MIM: 603637 MIM: 140750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
There are no transcripts associated with this gene.

Gene
ST5
Gene Name
suppression of tumorigenicity 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005418.3 3678 Missense Mutation NP_005409.3
NM_139157.2 3678 Missense Mutation NP_631896.1
NM_213618.1 3678 Missense Mutation NP_998783.1
XM_005253077.1 3678 Missense Mutation XP_005253134.1
XM_005253083.2 3678 Missense Mutation XP_005253140.1
XM_011520309.1 3678 Missense Mutation XP_011518611.1
XM_011520310.1 3678 Missense Mutation XP_011518612.1
XM_011520311.2 3678 Missense Mutation XP_011518613.1
XM_011520312.1 3678 Missense Mutation XP_011518614.1
XM_011520313.1 3678 Missense Mutation XP_011518615.1
XM_011520314.1 3678 Missense Mutation XP_011518616.1
XM_011520315.1 3678 Missense Mutation XP_011518617.1
XM_011520316.1 3678 Missense Mutation XP_011518618.1
XM_011520317.1 3678 Missense Mutation XP_011518619.1
XM_011520318.1 3678 Missense Mutation XP_011518620.1
XM_011520319.2 3678 Missense Mutation XP_011518621.1
XM_011520320.1 3678 Intron XP_011518622.1
XM_011520321.2 3678 Intron XP_011518623.1
XM_011520322.1 3678 Intron XP_011518624.1
XM_011520323.2 3678 Missense Mutation XP_011518625.2
XM_011520324.2 3678 Missense Mutation XP_011518626.2
XM_011520325.1 3678 Missense Mutation XP_011518627.1
XM_011520326.1 3678 Missense Mutation XP_011518628.1
XM_011520327.1 3678 Missense Mutation XP_011518629.1
XM_011520328.1 3678 Missense Mutation XP_011518630.1
XM_011520329.1 3678 Missense Mutation XP_011518631.1
XM_017018182.1 3678 Missense Mutation XP_016873671.1
XM_017018183.1 3678 Missense Mutation XP_016873672.1
XM_017018184.1 3678 Missense Mutation XP_016873673.1
XM_017018185.1 3678 Missense Mutation XP_016873674.1
XM_017018186.1 3678 Missense Mutation XP_016873675.1
XM_017018187.1 3678 Missense Mutation XP_016873676.1

View Full Product Details