Product Details
- SNP ID
-
rs137998609
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:617593 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGCCGTGGGGCTTCCGCCAGCTC[C/G]GGCCGCTGCGGGGGGCTCAGGGGCA
- Phenotype
-
MIM: 606839
MIM: 605047
MIM: 611780
MIM: 182099
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDHR5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34889521] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDHR5
- Gene Name
- cadherin related family member 5
- Gene
- IRF7
- Gene Name
- interferon regulatory factor 7
There are no transcripts associated with this gene.
- Gene
- PHRF1
- Gene Name
- PHD and ring finger domains 1
There are no transcripts associated with this gene.
- Gene
- SCT
- Gene Name
- secretin
There are no transcripts associated with this gene.
View Full Product Details