Product Details

SNP ID
rs138154701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112226460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGGAAGATGAGCACGGAAGGTG[G/T]TGGCCGTCGCTGCCAGGCACAAGTG
Phenotype
MIM: 611740 MIM: 612719
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCO2 PubMed Links

Gene Details

Gene
BCO2
Gene Name
beta-carotene oxygenase 2
There are no transcripts associated with this gene.

Gene
PTS
Gene Name
6-pyruvoyltetrahydropterin synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000317.2 96 Missense Mutation GGT,GTT G6V NP_000308.1

View Full Product Details