Product Details

SNP ID
rs138260730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5351967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATTGACAAGGGCTGGTCTGTCC[A/C]TTATGCCAATAGTTGTTCGCCTACA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 460 Intron NP_001005567.2
Gene
OR51B6
Gene Name
olfactory receptor family 51 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004750.1 460 Missense Mutation ATT,CTT I154L NP_001004750.1

View Full Product Details