Product Details

SNP ID
rs138374935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59422478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGATAGACAGAGTATGTTTCA[A/G]TGAAAGAACTAAGGAATCCACCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5A2 PubMed Links

Gene Details

Gene
OR5A2
Gene Name
olfactory receptor family 5 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001954.1 476 Missense Mutation ACT,ATT T159I NP_001001954.1

View Full Product Details