Product Details

SNP ID
rs138403273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4769266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGAATAATTAAGATATATGACA[C/G]GACAATGCATGGTGTATCTATTCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51F1 PubMed Links

Gene Details

Gene
OR51F1
Gene Name
olfactory receptor family 51 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004752.1 652 Silent Mutation CTG,GTG L218V NP_001004752.1

View Full Product Details