Product Details

SNP ID
rs138462032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9581550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTAGGTGGAAGTTTAGCTGATG[C/G]TATAAGTGAAAACTACAGAATCATG
Phenotype
MIM: 193525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WEE1 PubMed Links

Gene Details

Gene
WEE1
Gene Name
WEE1 G2 checkpoint kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143976.1 1273 Missense Mutation GCT,GGT A173G NP_001137448.1
NM_003390.3 1273 Missense Mutation GCT,GGT A387G NP_003381.1

View Full Product Details