Product Details

SNP ID
rs138530752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63463547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTATGCTATCCACTACAGCTG[A/T]AATAACTGCTCCAGCAGCCTTCGCT
Phenotype
MIM: 611474
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HRASLS5 PubMed Links

Gene Details

Gene
HRASLS5
Gene Name
HRAS like suppressor family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146728.1 794 Missense Mutation TAC,TTC Y249F NP_001140200.1
NM_001146729.1 794 Missense Mutation ACA,TCA T256S NP_001140201.1
NM_054108.3 794 Missense Mutation ACA,TCA T266S NP_473449.1
XM_011544751.2 794 Missense Mutation ACA,TCA T255S XP_011543053.1
XM_011544752.2 794 Missense Mutation ACA,TCA T154S XP_011543054.1
XM_011544753.1 794 Missense Mutation ACA,TCA T146S XP_011543055.1
XM_017017172.1 794 Missense Mutation ACA,TCA T146S XP_016872661.1

View Full Product Details