Product Details

SNP ID
rs138594141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110579079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGTTCCCATAAGTGCTTCCTCT[A/G]CAGAAGATGCTTTCTCTAGCCCTCT
Phenotype
MIM: 609568
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP20 PubMed Links

Gene Details

Gene
ARHGAP20
Gene Name
Rho GTPase activating protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258415.1 3973 UTR 3 NP_001245344.1
NM_001258416.1 3973 UTR 3 NP_001245345.1
NM_001258417.1 3973 UTR 3 NP_001245346.1
NM_001258418.1 3973 UTR 3 NP_001245347.1
NM_020809.3 3973 UTR 3 NP_065860.2
XM_005271628.3 3973 UTR 3 XP_005271685.1
XM_006718893.3 3973 UTR 3 XP_006718956.1

View Full Product Details