Product Details

SNP ID
rs138674997
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61300214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCGCTTCATACCGCTGCCATC[A/G]TCATACTGCAATGAGAAGATGGGCG
Phenotype
MIM: 600045 MIM: 611115
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DDB1 PubMed Links

Gene Details

Gene
DDB1
Gene Name
damage specific DNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001923.4 3589 Silent Mutation GAC,GAT D1115D NP_001914.3
Gene
VWCE
Gene Name
von Willebrand factor C and EGF domains
There are no transcripts associated with this gene.

View Full Product Details