Product Details
- SNP ID
-
rs138685890
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72000376 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTCAGCCCTTTGTGGGTCCTGCT[C/T]CTGTGTGCCCACGTCGTCACTCTCC
- Phenotype
-
MIM: 604113
MIM: 164009
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL18BP
PubMed Links
Gene Details
- Gene
- IL18BP
- Gene Name
- interleukin 18 binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039659.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_001034748.1 |
NM_001039660.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_001034749.1 |
NM_001145055.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_001138527.1 |
NM_001145057.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_001138529.1 |
NM_005699.3 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_005690.2 |
NM_173042.2 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_766630.2 |
NM_173044.2 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
NP_766632.2 |
XM_017017059.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
XP_016872548.1 |
XM_017017060.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
XP_016872549.1 |
XM_017017061.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
XP_016872550.1 |
XM_017017062.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
XP_016872551.1 |
XM_017017063.1 |
658 |
Silent Mutation |
CTC,CTT |
L18L |
XP_016872552.1 |
- Gene
- NUMA1
- Gene Name
- nuclear mitotic apparatus protein 1
There are no transcripts associated with this gene.
- Gene
- RNF121
- Gene Name
- ring finger protein 121
There are no transcripts associated with this gene.
View Full Product Details