Product Details

SNP ID
rs138717332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72293466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGATCTCCAGACGCAGCTTGGG[A/G]AGGCGCTTCTCAGCCTGGGGTGAGG
Phenotype
MIM: 616254
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPB PubMed Links

Gene Details

Gene
CLPB
Gene Name
ClpB homolog, mitochondrial AAA ATPase chaperonin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258392.2 2235 Silent Mutation CTC,CTT L645L NP_001245321.1
NM_001258393.2 2235 Silent Mutation CTC,CTT L616L NP_001245322.1
NM_001258394.2 2235 Silent Mutation CTC,CTT L630L NP_001245323.1
NM_030813.5 2235 Silent Mutation CTC,CTT L675L NP_110440.1
XM_005274320.1 2235 Silent Mutation CTC,CTT L646L XP_005274377.1
XM_011545289.2 2235 Intron XP_011543591.1
Gene
LOC105369375
Gene Name
uncharacterized LOC105369375
There are no transcripts associated with this gene.

View Full Product Details