Product Details

SNP ID
rs139123311
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65862462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACAGGTAGCCCCTGGGAGTGCT[C/T]GACCCTGGCCAGCCCTCCGCTCCCT
Phenotype
MIM: 601442 MIM: 604633 MIM: 606591
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CFL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs545500,rs659857] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CFL1
Gene Name
cofilin 1
There are no transcripts associated with this gene.

Gene
EFEMP2
Gene Name
EGF containing fibulin like extracellular matrix protein 2
There are no transcripts associated with this gene.

Gene
MUS81
Gene Name
MUS81 structure-specific endonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025128.4 706 Nonsense Mutation CGA,TGA R180* NP_079404.3
XM_005274307.2 706 Nonsense Mutation CGA,TGA R180* XP_005274364.1
XM_011545269.1 706 Nonsense Mutation CGA,TGA R180* XP_011543571.1
XM_011545270.1 706 Nonsense Mutation CGA,TGA R180* XP_011543572.1
Gene
SNX32
Gene Name
sorting nexin 32
There are no transcripts associated with this gene.

View Full Product Details