Product Details

SNP ID
rs139269006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65018424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTTCAATGGGGAGGACATCGA[C/T]ACCATCTCCCCAACGCTGGGCTTCA
Phenotype
MIM: 601175 MIM: 605964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL2 PubMed Links

Gene Details

Gene
ARL2
Gene Name
ADP ribosylation factor like GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199745.1 221 Silent Mutation GAC,GAT D42D NP_001186674.1
NM_001667.3 221 Silent Mutation GAC,GAT D42D NP_001658.2
Gene
ARL2-SNX15
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
MIR6879
Gene Name
microRNA 6879
There are no transcripts associated with this gene.

Gene
SNX15
Gene Name
sorting nexin 15
There are no transcripts associated with this gene.

View Full Product Details