Product Details

SNP ID
rs139274381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4572147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACATGACCTAATAGTTATGTCTC[A/C]ACACAAACCTGACCTGCTAACCAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf40 PubMed Links

Gene Details

Gene
C11orf40
Gene Name
chromosome 11 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144663.1 Intron NP_653264.1

View Full Product Details