Product Details

SNP ID
rs139281157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55839379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACTGTTGCCACTTTTAATGAG[A/C]TAAGCACACTACTCATCATTCTGAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5D16 PubMed Links

Gene Details

Gene
OR5D16
Gene Name
olfactory receptor family 5 subfamily D member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005496.1 628 Missense Mutation ATA,CTA I210L NP_001005496.1

View Full Product Details