Product Details

SNP ID
rs139345196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62575551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGCTGCCCCTCTTCTCCAGGG[C/G]CTCCATGCTTTCCAGTGGTCAGGGG
Phenotype
MIM: 130593 MIM: 610641
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EEF1G PubMed Links

Gene Details

Gene
EEF1G
Gene Name
eukaryotic translation elongation factor 1 gamma
There are no transcripts associated with this gene.

Gene
MIR6747
Gene Name
microRNA 6747
There are no transcripts associated with this gene.

Gene
TUT1
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022830.2 2320 Missense Mutation GCC,GGC A761G NP_073741.2

View Full Product Details