Product Details

SNP ID
rs139425981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2947798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCTCATGGGACCATTTAAAAG[A/G]TGGGGTTTGAACAGATAACACTCGA
Phenotype
MIM: 601651
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAP1L4 PubMed Links

Gene Details

Gene
NAP1L4
Gene Name
nucleosome assembly protein 1 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005969.3 Intron NP_005960.1

View Full Product Details