Product Details

SNP ID
rs139429864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6211703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGCGTTCAGGGGCCCCGCCCC[A/G]GGTGAGTAGAACTGGAGTTGAAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf42 PubMed Links

Gene Details

Gene
C11orf42
Gene Name
chromosome 11 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173525.2 3059 Intron NP_775796.2
XM_011519926.2 3059 Intron XP_011518228.1
XM_011519927.2 3059 Intron XP_011518229.1
Gene
FAM160A2
Gene Name
family with sequence similarity 160 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098794.1 3059 Silent Mutation NP_001092264.1
NM_032127.3 3059 Silent Mutation NP_115503.2
XM_006718343.2 3059 Missense Mutation XP_006718406.1
XM_011520397.2 3059 Missense Mutation XP_011518699.1
XM_011520398.2 3059 Intron XP_011518700.1
XM_011520399.2 3059 Intron XP_011518701.1

View Full Product Details