Product Details

SNP ID
rs139464903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33044021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGCCTCCTTGTGCATGTGGCC[A/G]TGAGCTAGGTTCTAATAATATAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCP11L1 PubMed Links

Gene Details

Gene
TCP11L1
Gene Name
t-complex 11 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145541.1 Intron NP_001139013.1
NM_018393.3 Intron NP_060863.3
XM_011520204.2 Intron XP_011518506.1
XM_011520205.2 Intron XP_011518507.1
XM_011520206.2 Intron XP_011518508.1
XM_011520207.2 Intron XP_011518509.1
XM_017017989.1 Intron XP_016873478.1
XM_017017990.1 Intron XP_016873479.1
XM_017017991.1 Intron XP_016873480.1
XM_017017992.1 Intron XP_016873481.1

View Full Product Details