Product Details

SNP ID
rs139540641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3856375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGAGAAGGCGACAGGAACCAG[C/T]TCGGGGGCCAACTCTGAGGAGTCCA
Phenotype
MIM: 605921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4687 PubMed Links

Gene Details

Gene
MIR4687
Gene Name
microRNA 4687
There are no transcripts associated with this gene.

Gene
STIM1
Gene Name
stromal interaction molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277961.1 673 Silent Mutation AGC,AGT S35S NP_001264890.1
NM_001277962.1 673 Silent Mutation AGC,AGT S35S NP_001264891.1
NM_003156.3 673 Silent Mutation AGC,AGT S35S NP_003147.2

View Full Product Details