Product Details

SNP ID
rs139641051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6846526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTTATCATTATCCTACACTGTT[A/G]TGACTCCCATGTTGAACCCCATTAT
Phenotype
MIM: 608493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10A5 PubMed Links

Gene Details

Gene
OR10A5
Gene Name
olfactory receptor family 10 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178168.1 844 Missense Mutation ATG,GTG M282V NP_835462.1

View Full Product Details