Product Details

SNP ID
rs139674102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47216408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCACAGATCCTGCCACCATGCC[A/G]CAGCATCGTCAGGACCCTCCACCAG
Phenotype
MIM: 600811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDB2 PubMed Links

Gene Details

Gene
DDB2
Gene Name
damage specific DNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000107.2 395 Missense Mutation CAC,CGC H67R NP_000098.1
NM_001300734.1 395 Missense Mutation CAC,CGC H67R NP_001287663.1

View Full Product Details