Product Details

SNP ID
rs139734641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6319311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCTTCAGCGCTCCGGCCAGGC[C/G]CAAGGCGAGGCGGCTTGACCGGGGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 672 Missense Mutation GCG,GGG A213G NP_659477.2

View Full Product Details