Product Details

SNP ID
rs139777859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4367571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGATGCAGACATGGGAGCCAAAT[A/G]TGTTGAGAGCTTTGTGGCAAGCATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52B4 PubMed Links

Gene Details

Gene
OR52B4
Gene Name
olfactory receptor family 52 subfamily B member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005161.3 816 Missense Mutation ACA,ATA T242I NP_001005161.2

View Full Product Details