Product Details

SNP ID
rs139850206
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126306632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTTGTGATCCTGGAGAAGACGCC[A/G]TTTCAGGTGGAACAGGTGGCTCAGC
Phenotype
MIM: 610534
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DCPS PubMed Links

Gene Details

Gene
DCPS
Gene Name
decapping enzyme, scavenger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014026.4 1044 Silent Mutation CCA,CCG P88P NP_054745.1
XM_011542778.2 1044 Silent Mutation CCA,CCG P95P XP_011541080.1
XM_011542780.2 1044 Intron XP_011541082.1
XM_017017612.1 1044 UTR 5 XP_016873101.1
Gene
HP11014
Gene Name
uncharacterized HP11014
There are no transcripts associated with this gene.

View Full Product Details