Product Details

SNP ID
rs139861196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34152438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCACGCGCTCTGCCAGGGTGTT[C/T]TGCAGGTCCTGCAGTGGATCCAGGC
Phenotype
MIM: 609221
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABTB2 PubMed Links

Gene Details

Gene
ABTB2
Gene Name
ankyrin repeat and BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145804.2 3452 Silent Mutation CAA,CAG Q1009Q NP_665803.2
Gene
NAT10
Gene Name
N-acetyltransferase 10
There are no transcripts associated with this gene.

View Full Product Details