Product Details

SNP ID
rs140082575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62243312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAATGCCGAGTTCTGCCCAGCT[C/G]TTGTTTCTGAGCTGTTAGACTTCTT
Phenotype
MIM: 615061
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCGB1D2 PubMed Links

Gene Details

Gene
SCGB1D2
Gene Name
secretoglobin family 1D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006551.3 135 Missense Mutation CTT,GTT L27V NP_006542.1

View Full Product Details