Product Details

SNP ID
rs140120221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5132036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGGATTGCAAAGGCAACAAATA[G/T]GCCATATATCTTGTTGACTCGGATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR52A5 PubMed Links

Gene Details

Gene
OR52A5
Gene Name
olfactory receptor family 52 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005160.2 607 Missense Mutation ATA,CTA I203L NP_001005160.1

View Full Product Details