Product Details

SNP ID
rs140279964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125445836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTAGCCAGACTACATAATGAGC[A/G]GTGAAAGCGGCTGCCTTCCCCTCTC
Phenotype
MIM: 604825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005103.4 1718 UTR 3 NP_005094.1
NM_022549.3 1718 Intron NP_072043.1
XM_005271734.2 1718 UTR 3 XP_005271791.1
XM_005271735.2 1718 UTR 3 XP_005271792.1

View Full Product Details