Product Details

SNP ID
rs140298838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47410328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCTCCTGGGTTCCCTCATGGT[A/G]GGGCTCAGTGGGGTCTTCCCGTTGC
Phenotype
MIM: 186852 MIM: 608735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4487 PubMed Links

Gene Details

Gene
MIR4487
Gene Name
microRNA 4487
There are no transcripts associated with this gene.

Gene
PSMC3
Gene Name
proteasome 26S subunit, ATPase 3
There are no transcripts associated with this gene.

Gene
SLC39A13
Gene Name
solute carrier family 39 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128225.2 289 Silent Mutation GTA,GTG V78V NP_001121697.1
NM_152264.4 289 Silent Mutation GTA,GTG V78V NP_689477.2
XM_006718381.3 289 Silent Mutation GTA,GTG V93V XP_006718444.1
XM_006718383.3 289 Silent Mutation GTA,GTG V93V XP_006718446.1
XM_006718384.2 289 Silent Mutation GTA,GTG V93V XP_006718447.1
XM_006718385.2 289 Silent Mutation GTA,GTG V93V XP_006718448.1
XM_011520466.1 289 Silent Mutation GTA,GTG V93V XP_011518768.1
XM_011520467.1 289 Silent Mutation GTA,GTG V78V XP_011518769.1
XM_011520468.2 289 Silent Mutation GTA,GTG V78V XP_011518770.1
XM_011520469.1 289 Silent Mutation GTA,GTG V93V XP_011518771.1
XM_011520470.1 289 Silent Mutation GTA,GTG V78V XP_011518772.1
XM_017018540.1 289 Silent Mutation GTA,GTG V78V XP_016874029.1
XM_017018541.1 289 Silent Mutation GTA,GTG V78V XP_016874030.1
XM_017018542.1 289 Silent Mutation GTA,GTG V78V XP_016874031.1

View Full Product Details