Product Details

SNP ID
rs140448421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55651003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTGGTGTTTTCTTTCTTCTAC[A/G]TAATCATTCTTCTGGGAAATCTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4S2 PubMed Links

Gene Details

Gene
OR4S2
Gene Name
olfactory receptor family 4 subfamily S member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004059.2 100 Missense Mutation ATA,GTA I34V NP_001004059.2

View Full Product Details