Product Details

SNP ID
rs140563330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22338580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGGCCCCAGGAAAAGAGGGGC[G/T]AAAGAATTTTGCTGGAAAATCACTC
Phenotype
MIM: 607563
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC17A6 PubMed Links

Gene Details

Gene
SLC17A6
Gene Name
solute carrier family 17 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020346.2 460 Missense Mutation CGA,CTA R16L NP_065079.1

View Full Product Details