Product Details
- SNP ID
-
rs140739403
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9021908 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTCCTGATGGTTCTCAGATGCAT[A/G]GAGCCTGCCATCTCGAACTATGTCT
- Phenotype
-
MIM: 611747
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCUBE2
PubMed Links
Gene Details
- Gene
- SCUBE2
- Gene Name
- signal peptide, CUB domain and EGF like domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170690.1 |
2973 |
Missense Mutation |
CAT,TAT |
H747Y |
NP_001164161.1 |
NM_020974.2 |
2973 |
Missense Mutation |
CAT,TAT |
H911Y |
NP_066025.2 |
XM_005253032.4 |
2973 |
Missense Mutation |
CAT,TAT |
H968Y |
XP_005253089.2 |
XM_005253033.4 |
2973 |
Missense Mutation |
CAT,TAT |
H939Y |
XP_005253090.2 |
XM_005253034.4 |
2973 |
Missense Mutation |
CAT,TAT |
H927Y |
XP_005253091.2 |
XM_005253035.4 |
2973 |
Intron |
|
|
XP_005253092.2 |
XM_005253036.4 |
2973 |
Missense Mutation |
CAT,TAT |
H859Y |
XP_005253093.2 |
XM_005253037.4 |
2973 |
Missense Mutation |
CAT,TAT |
H813Y |
XP_005253094.2 |
XM_011520246.2 |
2973 |
Missense Mutation |
CAT,TAT |
H902Y |
XP_011518548.1 |
XM_011520247.2 |
2973 |
Missense Mutation |
CAT,TAT |
H900Y |
XP_011518549.1 |
XM_011520248.2 |
2973 |
Intron |
|
|
XP_011518550.1 |
XM_017018080.1 |
2973 |
Missense Mutation |
CAT,TAT |
H898Y |
XP_016873569.1 |
XM_017018081.1 |
2973 |
Missense Mutation |
CAT,TAT |
H832Y |
XP_016873570.1 |
XM_017018082.1 |
2973 |
Intron |
|
|
XP_016873571.1 |
View Full Product Details