Product Details

SNP ID
rs140831932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65856198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGACTTACGCACCTTCATGTC[A/G]TTGAACACCTTGATGACACCATCAG
Phenotype
MIM: 601442 MIM: 606591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFL1 PubMed Links

Gene Details

Gene
CFL1
Gene Name
cofilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005507.2 282 Silent Mutation AAC,AAT N16N NP_005498.1
Gene
MUS81
Gene Name
MUS81 structure-specific endonuclease subunit
There are no transcripts associated with this gene.

Gene
SNX32
Gene Name
sorting nexin 32
There are no transcripts associated with this gene.

View Full Product Details