Product Details

SNP ID
rs140903243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134153182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACATAAGATGTCATTTCCTTGA[C/T]TCCGGCCTTCAATTTTCTCTAAAAG
Phenotype
MIM: 606871 MIM: 609276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JAM3 PubMed Links

Gene Details

Gene
JAM3
Gene Name
junctional adhesion molecule 3
There are no transcripts associated with this gene.

Gene
NCAPD3
Gene Name
non-SMC condensin II complex subunit D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015261.2 4952 Missense Mutation AAT,AGT N1449S NP_056076.1
XM_011542719.2 4952 Missense Mutation AAT,AGT N1311S XP_011541021.1

View Full Product Details