Product Details

SNP ID
rs141005361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2885244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACGCGGGCACCGAGTCGCTGTC[C/T]ACTTCGGTCCACTGCAGGCGTCCAG
Phenotype
MIM: 600856 MIM: 603240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN1C PubMed Links

Gene Details

Gene
CDKN1C
Gene Name
cyclin dependent kinase inhibitor 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000076.2 384 Silent Mutation GTA,GTG V82V NP_000067.1
NM_001122630.1 384 Silent Mutation GTA,GTG V71V NP_001116102.1
NM_001122631.1 384 Silent Mutation GTA,GTG V71V NP_001116103.1
XM_005252732.4 384 Silent Mutation GTA,GTG V71V XP_005252789.1
XM_017017088.1 384 Silent Mutation GTA,GTG V82V XP_016872577.1
Gene
SLC22A18AS
Gene Name
solute carrier family 22 member 18 antisense
There are no transcripts associated with this gene.

View Full Product Details