Product Details
- SNP ID
-
rs141150956
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59806527 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCGCATCTTCTCTAGAGTCCC[A/G]CGGCTCACAGCCTTTGCTGCGAAGG
- Phenotype
-
MIM: 611829
MIM: 600876
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147545257] are located under a probe and SNP(s) [rs491671] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL16
- Gene Name
- mitochondrial ribosomal protein L16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017840.3 |
790 |
Silent Mutation |
CGC,CGT |
R192R |
NP_060310.1 |
- Gene
- STX3
- Gene Name
- syntaxin 3
View Full Product Details