Product Details

SNP ID
rs141188154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75720170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAAGGACTATTTCCCCATCTC[A/G]GTGAGTATTGAGGCTGGTTGGGGTT
Phenotype
MIM: 610270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOGAT2 PubMed Links

Gene Details

Gene
MOGAT2
Gene Name
monoacylglycerol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025098.3 340 Silent Mutation TCA,TCG S90S NP_079374.2
XM_011545267.1 340 Silent Mutation TCA,TCG S90S XP_011543569.1

View Full Product Details