Product Details

SNP ID
rs141443392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59515102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACGCCCATGTACTTCCTGCTC[C/T]GCAACCTGTCTATTCTTGACATCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4D9 PubMed Links

Gene Details

Gene
OR4D9
Gene Name
olfactory receptor family 4 subfamily D member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004711.1 190 Missense Mutation CGC,TGC R64C NP_001004711.1

View Full Product Details