Product Details

SNP ID
rs141469388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6199609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCATGGCCTGTGATCGTGCTG[C/T]GGCAATAGGGCGTCCACTGCACTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf42 PubMed Links

Gene Details

Gene
C11orf42
Gene Name
chromosome 11 open reading frame 42
There are no transcripts associated with this gene.

Gene
OR52W1
Gene Name
olfactory receptor family 52 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005178.1 386 Missense Mutation GCG,GTG A129V NP_001005178.1

View Full Product Details