Product Details

SNP ID
rs141507953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55993834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTCCTGATTCAGGGAGTAGCT[C/G]GAACTAGGTCTCAGGTAAGTATAGA
Phenotype
MIM: 608492
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5F1 PubMed Links

Gene Details

Gene
OR5F1
Gene Name
olfactory receptor family 5 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003697.1 792 Silent Mutation TCC,TCG S264S NP_003688.1
Gene
OR7E5P
Gene Name
olfactory receptor family 7 subfamily E member 5 pseudogene
There are no transcripts associated with this gene.

View Full Product Details