Product Details

SNP ID
rs141513598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35663159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCGAGGAACTGCCTCACGACGG[C/G]ACGTGTGACGAGTGCGAGCCCGACG
Phenotype
MIM: 612298
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM44 PubMed Links

Gene Details

Gene
TRIM44
Gene Name
tripartite motif containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017583.5 408 Silent Mutation GGC,GGG G16G NP_060053.2
XM_006718254.1 408 Silent Mutation GGC,GGG G16G XP_006718317.1

View Full Product Details