Product Details

SNP ID
rs141564627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64827125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGTGGTTGAAGTTGGTAGGCGG[C/G]GAGATGAGCTTGGAGCGCACAAAAG
Phenotype
MIM: 613991
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017525.2 4463 Silent Mutation TCC,TCG S1438S NP_059995.2
XM_011545155.2 4463 Silent Mutation TCC,TCG S1456S XP_011543457.1
XM_011545156.2 4463 Silent Mutation TCC,TCG S1449S XP_011543458.1
XM_011545157.2 4463 Missense Mutation CCC,GCC P1424A XP_011543459.1
XM_011545158.2 4463 Missense Mutation CCC,GCC P1424A XP_011543460.1
XM_011545159.2 4463 Silent Mutation TCC,TCG S1456S XP_011543461.1
XM_011545160.2 4463 Silent Mutation TCC,TCG S1358S XP_011543462.1
XM_011545161.2 4463 Intron XP_011543463.1
XM_017017996.1 4463 Silent Mutation TCC,TCG S1455S XP_016873485.1
XM_017017997.1 4463 Intron XP_016873486.1
XM_017017998.1 4463 Intron XP_016873487.1
XM_017017999.1 4463 Silent Mutation TCC,TCG S752S XP_016873488.1

View Full Product Details