Product Details

SNP ID
rs141842162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106051236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTGTTTTCAAAGCCACACACA[C/T]ACAAACAAAAGCCTCCACTGAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD3 PubMed Links

Gene Details

Gene
KBTBD3
Gene Name
kelch repeat and BTB domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152433.3 3892 UTR 3 NP_689646.2
NM_198439.2 3892 UTR 3 NP_940841.1
XM_006718767.3 3892 UTR 3 XP_006718830.1
XM_011542618.2 3892 UTR 3 XP_011540920.1
XM_011542619.2 3892 UTR 3 XP_011540921.1

View Full Product Details