Product Details
- SNP ID
-
rs141898388
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:105029158 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAGCAGAAAGCGATAAAATCCTT[C/T]TCTATGTGGGCTTTCTTAATAGCAT
- Phenotype
-
MIM: 147678
MIM: 602665
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CASP1
PubMed Links
Gene Details
- Gene
- CASP1
- Gene Name
- caspase 1
- Gene
- CASP5
- Gene Name
- caspase 5
There are no transcripts associated with this gene.
View Full Product Details